4,6-Dichloropyridine-3-carbaldehyde

Supplier: Apollo Scientific
Warning

Synonyms: 4,6-Dichloronicotinaldehyde

OR303638-1G OR303638-5G
APOSOR303638-1GEA 82.9 EUR
APOSOR303638-1G APOSOR303638-5G
4,6-Dichloropyridine-3-carbaldehyde
4,6-Dichloropyridine-3-carbaldehyde
Formula: C₆H₃Cl₂NO
MW: 176 g/mol
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD12025485
CAS Number: 1060811-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR