4,6-Dichloronicotinonitrile 98%

Supplier: Apollo Scientific
Warning

OR303715-25G
APOSOR303715-25GEA 87 EUR
APOSOR303715-25G
4,6-Dichloronicotinonitrile 98%
4,6-Dichloronicotinonitrile
Formula: C₆H₂Cl₂N₂
Melting Pt: 133…135 °C
Storage Temperature: Ambient
MDL Number: MFCD08276124
CAS Number: 166526-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR