3-(Benzyloxy)-5-bromo-2-methyl-4-pyridinol

Supplier: Apollo Scientific

OR303719-1G OR303719-500MG
APOSOR303719-1GEA 364 EUR
APOSOR303719-1G APOSOR303719-500MG
3-(Benzyloxy)-5-bromo-2-methyl-4-pyridinol
3-(Benzyloxy)-5-bromo-2-methyl-4-pyridinol
Formula: C₁₃H₁₂BrNO₂
Melting Pt: 197…199 °C
Storage Temperature: Ambient
MDL Number: MFCD16140264
CAS Number: 906657-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR