Methyl-6-formylnicotinate 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 6-formylnicotinate
OR303729-1G
OR303729-5G
APOSOR303729-1GEA
101
EUR
APOSOR303729-1G
APOSOR303729-5G
Methyl-6-formylnicotinate 97%
Methyl-6-formylnicotinate
Formula:
C₈H₇NO₃ MW: 165.1500 g/mol Melting Pt: 115…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD08275010 CAS Number: 10165-86-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...