1-(2,6-Dichloropyridin-3-yl)ethanone

Supplier: Apollo Scientific
Warning

Synonyms: 3-Acetyl-2,6-dichloropyridine

OR303734-1G OR303734-5G
APOSOR303734-1GEA 88.6 EUR
APOSOR303734-1G APOSOR303734-5G
1-(2,6-Dichloropyridin-3-yl)ethanone
1-(2,6-Dichloropyridin-3-yl)ethanone
Formula: C₇H₅Cl₂NO
MW: 190,0267 g/mol
Melting Pt: 32…34 °C
Storage Temperature: Ambient
MDL Number: MFCD11847148
CAS Number: 412018-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR