Ethyl-5-(cyanomethyl)-2-pyridinecarboxylate

Supplier: Apollo Scientific
Warning

OR303735-1G OR303735-500MG
APOSOR303735-1GEA 364 EUR
APOSOR303735-1G APOSOR303735-500MG
Ethyl-5-(cyanomethyl)-2-pyridinecarboxylate
Ethyl-5-(cyanomethyl)-2-pyridinecarboxylate
Formula: C₁₀H₁₀N₂O₂
MW: 190,2 g/mol
Boiling Pt: 160 °C/0,01mmHg
Storage Temperature: Ambient
MDL Number: MFCD16140285
CAS Number: 913839-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR