5-Chloro-2-pyridinecarbohydrazide 95%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Chloropicolinohydrazide

OR303749-1G OR303749-500MG
APOSOR303749-1GEA 132 EUR
APOSOR303749-1G APOSOR303749-500MG
5-Chloro-2-pyridinecarbohydrazide 95%
5-Chloro-2-pyridinecarbohydrazide
Formula: C₆H₆ClN₃O
MW: 171,59 g/mol
Melting Pt: 178…180 °C
Storage Temperature: Ambient
MDL Number: MFCD16622817
CAS Number: 145835-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR