tert-Butyl-4-(6-chloropyridin-3-yl)piperazine-1-carboxylate 95%
Supplier: Apollo Scientific
OR303754-1G
OR303754-500MG
APOSOR303754-1GEA
538
EUR
APOSOR303754-1G
APOSOR303754-500MG
tert-Butyl-4-(6-chloropyridin-3-yl)piperazine-1-carboxylate 95%
tert-Butyl-4-(6-chloropyridin-3-yl)piperazine-1-carboxylate
Formula:
C₁₄H₂₀ClN₃O₂ MW: 297,79 g/mol Melting Pt: 118…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD12923074 CAS Number: 633283-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...