5-Chloro-3-ethynyl-2-pyridinamine

Supplier: Apollo Scientific
Warning

Synonyms: 2-Amino-5-chloro-3-ethynylpyridine

OR303787-250MG OR303787-100MG
APOSOR303787-250MGEA 430 EUR
APOSOR303787-250MG APOSOR303787-100MG
5-Chloro-3-ethynyl-2-pyridinamine
5-Chloro-3-ethynyl-2-pyridinamine
Formula: C₇H₅ClN₂
MW: 152,58 g/mol
Melting Pt: 118…119 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10758079
CAS Number: 866318-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR