Methyl-4-aminopicolinate 97%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Aminopyridine-2-carboxylic acid methyll ester, Methyl 4-Aminopyridine-2-carboxylate, 4-Aminopicolinic acid methyll ester, Methyl 4-Aminopicolinate

OR303823-1G OR303823-5G OR303823-25G
APOSOR303823-1GEA 71.3 EUR
APOSOR303823-1G APOSOR303823-5G APOSOR303823-25G
Methyl-4-aminopicolinate 97%
Methyl 4-aminopicolinate
Formula: C₇H₈N₂O₂
MW: 152,1500 g/mol
Melting Pt: 129…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00956000
CAS Number: 71469-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR