5-Bromo-2-nitrobenzaldehyde

Supplier: Apollo Scientific
Warning

OR303921-5G OR303921-10G OR303921-25G
APOSOR303921-5GEA 21.9 EUR
APOSOR303921-5G APOSOR303921-10G APOSOR303921-25G
5-Bromo-2-nitrobenzaldehyde
5-Bromo-2-nitrobenzaldehyde
Formula: C₇H₄BrNO₃
Melting Pt: 70…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00456508
CAS Number: 20357-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR