4-Chloro-7-methyl-7H-pyrrolo[2,3-d]pyrimidine

Supplier: Apollo Scientific

OR303930-1G OR303930-5G OR303930-10G
APOSOR303930-1GEA 91.6 EUR
APOSOR303930-1G APOSOR303930-5G APOSOR303930-10G
4-Chloro-7-methyl-7H-pyrrolo[2,3-d]pyrimidine
4-Chloro-7-methyl-7H-pyrrolo[2,3-d]pyrimidine
Formula: C₇H₆ClN₃
MW: 167,6 g/mol
Melting Pt: 125…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00619245
CAS Number: 7781-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR