9,10-Dibromoanthracene

Supplier: Apollo Scientific
Warning

OR30417-100G OR30417-25G
APOSOR30417-100GEA 109 EUR
APOSOR30417-100G APOSOR30417-25G
9,10-Dibromoanthracene
9,10-Dibromoanthracene
Formula: C₁₄H₈Br₂
Melting Pt: 222…224 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00001244
CAS Number: 523-27-3
EINECS: 208-342-4
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR