1-Amino-4-naphthol hydrochloride 95%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Amino-1-naphthol hydrochloride

OR30442-25G OR30442-5G
APOSOR30442-25GEA 197 EUR
APOSOR30442-25G APOSOR30442-5G
1-Amino-4-naphthol hydrochloride 95%
1-Amino-4-naphthol hydrochloride
Formula: C₁₀H₁₀ClNO
MW: 195,65 g/mol
Melting Pt: 272…273 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012562
CAS Number: 5959-56-8
EINECS: 227-727-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR