2-Chloro-5-nitropyrimidine 95%

Supplier: Apollo Scientific
Danger

OR30486-5G OR30486-1G
APOSOR30486-5GEA 75.4 EUR
APOSOR30486-5G APOSOR30486-1G
2-Chloro-5-nitropyrimidine 95%
2-Chloro-5-nitropyrimidine
Formula: C₄H₂ClN₃O₂
MW: 159,53 g/mol
Melting Pt: 105…110 °C
Storage Temperature: Freezer
MDL Number: MFCD04117995
CAS Number: 10320-42-0
EINECS: 233-703-8
UN: 3335
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR