2-Indanone 98%

Supplier: Apollo Scientific
Warning

Synonyms: beta-Hydrindone, 2-Oxoindane, 1H-Inden-2(3H)-one

OR30508-100G OR30508-25G OR30508-5G
APOSOR30508-100GEA 88.6 EUR
APOSOR30508-100G APOSOR30508-25G APOSOR30508-5G
2-Indanone 98%
2-Indanone
Formula: C₉H₈O
MW: 132,16 g/mol
Melting Pt: 51…54 °C
Flash Pt: 100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003792
CAS Number: 615-13-4
EINECS: 210-410-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR