6-Bromo-m-toluic acid 98%
Supplier: Apollo Scientific
|
Warning
|
OR3053-1G
OR3053-5G
APOSOR3053-1GEA
21.9
EUR
APOSOR3053-1G
APOSOR3053-5G
6-Bromo-m-toluic acid 98%
6-Bromo-m-toluic acid
Formula:
C₈H₇BrO₂ MW: 215,05 g/mol Melting Pt: 136…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00079722 CAS Number: 6967-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...