Ethyl-5-oxo-4,5-dihydroisoxazole-4-carboxylate

Supplier: Apollo Scientific
Warning

OR305307-1G OR305307-5G
APOSOR305307-1GEA 654 EUR
APOSOR305307-1G APOSOR305307-5G
Ethyl-5-oxo-4,5-dihydroisoxazole-4-carboxylate
Ethyl-5-oxo-4,5-dihydroisoxazole-4-carboxylate
Formula: C₆H₇NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00020816
CAS Number: 500348-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR