5-Pyrimidinecarbonyl chloride

Supplier: Apollo Scientific
Danger

OR305377-1G OR305377-5G
APOSOR305377-1GEA 241 EUR
APOSOR305377-1G APOSOR305377-5G
5-Pyrimidinecarbonyl chloride
5-Pyrimidinecarbonyl chloride
Formula: C₅H₃ClN₂O
MW: 142,55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03789627
CAS Number: 40929-48-4
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR