2-Acetamido-6-methylpyridine
Supplier: Apollo Scientific
Synonyms:
N-(6-Methylpyridin-2-yl)acetamide
OR305411-10G
OR305411-5G
APOSOR305411-10GEA
97.4
EUR
APOSOR305411-10G
APOSOR305411-5G
2-Acetamido-6-methylpyridine
2-Acetamido-6-methylpyridine
Formula:
C₈H₁₀N₂O MW: 150,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00091881 CAS Number: 5327-33-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...