3-Bromo-o-toluic acid

Supplier: Apollo Scientific
Warning

OR3055-100G OR3055-25G
APOSOR3055-100GEA 85.7 EUR
APOSOR3055-100G APOSOR3055-25G
3-Bromo-o-toluic acid
3-Bromo-o-toluic acid
Formula: C₈H₇BrO₂
MW: 215,05 g/mol
Melting Pt: 152…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00270097
CAS Number: 76006-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR