1,4-Dibromonaphthalene 98%

Supplier: Apollo Scientific
Danger

OR30594-25G OR30594-5G
APOSOR30594-25GEA 84.4 EUR
APOSOR30594-25G APOSOR30594-5G
1,4-Dibromonaphthalene 98%
1,4-Dibromonaphthalene
Formula: C₁₀H₆Br₂
MW: 285,97 g/mol
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00041823
CAS Number: 83-53-4
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR