2-Naphthyl boronic acid pinacol ester

Supplier: Apollo Scientific
Warning

OR30614-1G OR30614-5G
APOSOR30614-1GEA 52.4 EUR
APOSOR30614-1G APOSOR30614-5G
2-Naphthyl boronic acid pinacol ester
2-Naphthyl boronic acid pinacol ester
Formula: C₁₆H₁₉BO₂
Storage Temperature: Ambient
MDL Number: MFCD05663868
CAS Number: 256652-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR