1-Aminopiperidine 97%

Supplier: Apollo Scientific
Danger

OR30621-25G OR30621-100G
APOSOR30621-25GEA 255 EUR
APOSOR30621-25G APOSOR30621-100G
1-Aminopiperidine 97%
1-Aminopiperidine
Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 145…147 °C/760mmHg
Density: 0.928 g/cm³ (25 °C)
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00006489
CAS Number: 2213-43-6
UN: 2920
ADR: 8+3 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR