1,18-Octadecanedioic acid 98%

Supplier: Apollo Scientific
Warning

OR30639-1G OR30639-5G
APOSOR30639-1GEA 21.9 EUR
APOSOR30639-1G APOSOR30639-5G
1,18-Octadecanedioic acid 98%
1,18-Octadecanedioic acid
Formula: C₁₈H₃₄O₄
Melting Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD00142369
CAS Number: 871-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR