2-Bromoethanol 97%

Supplier: Apollo Scientific
Danger

OR30640-100G OR30640-250G
APOSOR30640-100GEA 63.8 EUR
APOSOR30640-100G APOSOR30640-250G
2-Bromoethanol 97%
2-Bromoethanol
Formula: BrH₂CCH₂OH
Boiling Pt: 149…150 °C
Melting Pt: –80 °C
Density: 1,762 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002827
CAS Number: 540-51-2
EINECS: 208-748-1
UN: 2922
ADR: 8+6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR