6-Oxaspiro[2.5]octane-1-carboxylic acid

Supplier: Apollo Scientific

OR306524-100MG OR306524-1G OR306524-250MG
APOSOR306524-100MGEA 106 EUR
APOSOR306524-100MG APOSOR306524-1G APOSOR306524-250MG
6-Oxaspiro[2.5]octane-1-carboxylic acid
6-Oxaspiro[2.5]octane-1-carboxylic acid
Formula: C₈H₁₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD16744613
CAS Number: 909406-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR