1-(2-(Methylsulphonyl)phenyl)piperazine hydrochloride
Supplier: Apollo Scientific
|
Warning
|
OR30665-1G
OR30665-250MG
APOSOR30665-1GEA
79.9
EUR
APOSOR30665-1G
APOSOR30665-250MG
1-(2-(Methylsulphonyl)phenyl)piperazine hydrochloride
1-(2-(Methylsulphonyl)phenyl)piperazine hydrochloride
Formula:
C₁₁H₁₇ClN₂O₂S MW: 276.7800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11869755 CAS Number: 916488-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...