1-Bromo-4-pentylbenzene
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Pentylbromobenzene, 4-Bromopentylbenzene, (4-Bromophenyl)pentane
OR3070-100G
OR3070-10G
OR3070-25G
APOSOR3070-100GEA
58
EUR
APOSOR3070-100G
APOSOR3070-10G
APOSOR3070-25G
1-Bromo-4-pentylbenzene
1-Bromo-4-pentylbenzene
Formula:
C₁₁H₁₅Br MW: 227,14 g/mol Boiling Pt: 203…204 °C Density: 1,272 g/cm³ (25 °C) Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061113 CAS Number: 51554-95-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...