2-Methoxy-5-pyridineboronic acid pinacol ester
Supplier: Apollo Scientific
|
Warning
|
OR307018-1G
OR307018-25G
OR307018-5G
APOSOR307018-1GEA
58.2
EUR
APOSOR307018-1G
APOSOR307018-25G
APOSOR307018-5G
2-Methoxy-5-pyridineboronic acid pinacol ester
2-Methoxy-5-pyridineboronic acid pinacol ester
Formula:
C₁₂H₁₈BNO₃ MW: 235,09 g/mol Melting Pt: 47…51 °C Flash Pt: 102,2 °C Storage Temperature: Ambient |
MDL Number:
MFCD05663858 CAS Number: 445264-61-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...