4-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid 98%
Supplier: Apollo Scientific
|
Warning
|
OR307034-1G
OR307034-25G
OR307034-5G
APOSOR307034-1GEA
14.7
EUR
APOSOR307034-1G
APOSOR307034-25G
APOSOR307034-5G
4-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid 98%
4-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid
Formula:
C₁₀H₁₈N₂O₄ Melting Pt: 223 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD01076218 CAS Number: 128019-59-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...