N-Benzyltropinone
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
8-Benzyl-8-azabicyclo[3.2.1]octan-3-one, (1R,5S)-8-Benzyl-8-azabicyclo[3.2.1]octan-3-one
OR30704-1G
OR30704-25G
OR30704-5G
APOSOR30704-1GEA
21.9
EUR
APOSOR30704-1G
APOSOR30704-25G
APOSOR30704-5G
N-Benzyltropinone
N-Benzyltropinone
Formula:
C₁₄H₁₇NO MW: 215,30 g/mol Boiling Pt: 124 °C/0,2mmHg Melting Pt: 28…31 °C Density: 1,11 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00151640 CAS Number: 28957-72-4 EINECS: 249-329-3 UN: 2735 ADR: 8,III |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...