(S)-1-Boc-pyrrolidine-3-carboxylic acid

Supplier: Apollo Scientific

Synonyms: (S)-1-Boc-Pyrrolidine-3-carboxylic acid, (3S)-1-tert-butoxycarbonylpyrrolidine-3-carboxylic acid

OR307105-25G OR307105-5G
APOSOR307105-25GEA 362 EUR
APOSOR307105-25G APOSOR307105-5G
(S)-1-Boc-pyrrolidine-3-carboxylic acid
(S)-1-Boc-pyrrolidine-3-carboxylic acid
Formula: C₁₀H₁₇NO₄
MW: 215,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094728
CAS Number: 140148-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR