2-Methyl-2H-indazole-6-carboxylic acid methyl ester
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 2-methyl-2H-indazole-6-carboxylate
OR30724-1G
OR30724-5G
OR30724-10G
APOSOR30724-1GEA
50.9
EUR
APOSOR30724-1G
APOSOR30724-5G
APOSOR30724-10G
2-Methyl-2H-indazole-6-carboxylic acid methyl ester
2-Methyl-2H-indazole-6-carboxylic acid methyl ester
Formula:
C₁₀H₁₀N₂O₂ MW: 190,2 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11109407 CAS Number: 1071433-01-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...