4-Phenylpyridine-N-oxide 98%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Phenylpyridin-1-ium-1-olate, 4-Phenylpyridine 1-oxide

OR307240-5G OR307240-1G OR307240-25G
APOSOR307240-5GEA 156 EUR
APOSOR307240-5G APOSOR307240-1G APOSOR307240-25G
4-Phenylpyridine-N-oxide 98%
4-Phenylpyridine N-oxide
Formula: C₁₁H₉NO
MW: 171.1953 g/mol
Melting Pt: 153…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00006208
CAS Number: 1131-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR