3-Amino-3-(2-thienyl)propanoic acid

Supplier: Apollo Scientific
Warning

OR307538-1G OR307538-10G OR307538-5G
APOSOR307538-1GEA 23.2 EUR
APOSOR307538-1G APOSOR307538-10G APOSOR307538-5G
3-Amino-3-(2-thienyl)propanoic acid
3-Amino-3-(2-thienyl)propanoic acid
Formula: C₇H₉NO₂S
MW: 171,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00152178
CAS Number: 18389-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR