2-Chloro-6-methylquinoline-3-carbonitrile

Supplier: Apollo Scientific

OR307828-1G OR307828-5G
APOSOR307828-1GEA 293 EUR
APOSOR307828-1G APOSOR307828-5G
2-Chloro-6-methylquinoline-3-carbonitrile
2-Chloro-6-methylquinoline-3-carbonitrile
Formula: C₁₁H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD03045343
CAS Number: 483287-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR