2-Chloro-8-methylquinoline-3-carbonitrile

Supplier: Apollo Scientific

OR307833-1G OR307833-5G
APOSOR307833-1GEA 219 EUR
APOSOR307833-1G APOSOR307833-5G
2-Chloro-8-methylquinoline-3-carbonitrile
2-Chloro-8-methylquinoline-3-carbonitrile
Formula: C₁₁H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD08437565
CAS Number: 136812-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR