2-Chloro-3-cyanoquinoline  

Supplier: Apollo Scientific

Synonyms: 2-Chloroquinoline-3-carbonitrile

OR307835-1G OR307835-5G
APOSOR307835-1GEA 293 EUR
APOSOR307835-1G APOSOR307835-5G
2-Chloro-3-cyanoquinoline  
2-Chloro-3-cyanoquinoline
Formula: C₁₀H₅ClN₂
MW: 188,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08437566
CAS Number: 95104-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR