4-Amino-2,8-dimethylquinoline
Supplier: Apollo Scientific
OR307859-1G
OR307859-5G
APOSOR307859-1GEA
335
EUR
APOSOR307859-1G
APOSOR307859-5G
4-Amino-2,8-dimethylquinoline
4-Amino-2,8-dimethylquinoline
Formula:
C₁₁H₁₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD08669719 CAS Number: 51617-12-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...