2-Chloro-3,7-dimethylquinoline 98%
Supplier: Apollo Scientific
OR307902-1G
OR307902-5G
APOSOR307902-1GEA
285
EUR
APOSOR307902-1G
APOSOR307902-5G
2-Chloro-3,7-dimethylquinoline 98%
2-Chloro-3,7-dimethylquinoline
Formula:
C₁₁H₁₀ClN Storage Temperature: Ambient |
CAS Number:
73863-46-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...