2-Chloro-3-(chloromethyl)quinoline

Supplier: Apollo Scientific
Danger

OR307910-5G
APOSOR307910-5GEA 95.7 EUR
APOSOR307910-5G
2-Chloro-3-(chloromethyl)quinoline
2-Chloro-3-(chloromethyl)quinoline
Formula: C₁₀H₇Cl₂N
MW: 212,0753 g/mol
Melting Pt: 117 °C
Storage Temperature: Ambient
MDL Number: MFCD05865134
CAS Number: 90097-52-2
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR