6,7-Dimethylquinoline-2,3-dicarboxylic acid

Supplier: Apollo Scientific

OR307997-1G OR307997-5G
APOSOR307997-1GEA 310 EUR
APOSOR307997-1G APOSOR307997-5G
6,7-Dimethylquinoline-2,3-dicarboxylic acid
6,7-Dimethylquinoline-2,3-dicarboxylic acid
Formula: C₁₃H₁₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD09787799
CAS Number: 948294-45-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR