3-Iodooxetane
Supplier: Apollo Scientific
|
Warning
|
OR308004-1G
OR308004-5G
APOSOR308004-1GEA
21.9
EUR
APOSOR308004-1G
APOSOR308004-5G
3-Iodooxetane
3-Iodooxetane
Formula:
C₃H₅IO Boiling Pt: 166 °C Density: 2,14 g/cm³ (25 °C) Flash Pt: 54 °C Storage Temperature: Ambient |
MDL Number:
MFCD08544403 CAS Number: 26272-85-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...