3-Amino-3-methyloxetane 97%

Supplier: Apollo Scientific

Synonyms: 3-Methyloxetan-3-amine, 3-Methyl-3-oxetanamine

OR308009-1G OR308009-5G
APOSOR308009-1GEA 111 EUR
APOSOR308009-1G APOSOR308009-5G
3-Amino-3-methyloxetane 97%
3-Amino-3-methyloxetane
Formula: C₄H₉NO
MW: 87,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111713
CAS Number: 874473-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR