Methyl azetidine-3-carboxylate hydrochloride
Supplier: Apollo Scientific
|
Warning
|
OR308020-100G
OR308020-25G
OR308020-5G
APOSOR308020-100GEA
351
EUR
APOSOR308020-100G
APOSOR308020-25G
APOSOR308020-5G
Methyl azetidine-3-carboxylate hydrochloride
Methyl azetidine-3-carboxylate hydrochloride
Formula:
C₅H₁₀ClNO₂ Boiling Pt: 187…192 °C Storage Temperature: Ambient |
MDL Number:
MFCD01861758 CAS Number: 100202-39-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...