1-Boc-piperazine-2-carboxylic acid

Supplier: Apollo Scientific

Synonyms: 1-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid

OR308063-1G OR308063-5G
APOSOR308063-1GEA 72.7 EUR
APOSOR308063-1G APOSOR308063-5G
1-Boc-piperazine-2-carboxylic acid
1-Boc-piperazine-2-carboxylic acid
Formula: C₁₀H₁₈N₂O₄
MW: 230,2600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02682403
CAS Number: 1214196-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR