(S)-1-Boc-3-(hydroxymethyl)piperazine
Supplier: Apollo Scientific
OR308072-1G
OR308072-5G
APOSOR308072-1GEA
71.3
EUR
APOSOR308072-1G
APOSOR308072-5G
(S)-1-Boc-3-(hydroxymethyl)piperazine
(S)-1-Boc-3-(hydroxymethyl)piperazine
Formula:
C₁₀H₂₀N₂O₃ MW: 216,2800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05863889 CAS Number: 314741-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...