4-Methylpiperidin-4-ol
Supplier: Apollo Scientific
|
Danger
|
OR308124-5G
APOSOR308124-5GEA
82.8
EUR
APOSOR308124-5G
4-Methylpiperidin-4-ol
4-Methylpiperidin-4-ol
Formula:
C₆H₁₃NO Storage Temperature: Ambient |
MDL Number:
MFCD11100993 CAS Number: 3970-68-1 UN: 3263 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...