tert-Butyl methyl(piperidin-3-yl)carbamate 97%

Supplier: Apollo Scientific
Warning

OR308128-1G OR308128-5G
APOSOR308128-1GEA 78.3 EUR
APOSOR308128-1G APOSOR308128-5G
tert-Butyl methyl(piperidin-3-yl)carbamate 97%
tert-Butyl methyl(piperidin-3-yl)carbamate
Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06656642
CAS Number: 172478-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR